Plastic Graduated Beakers
: Wards
Autoclavable, microwavable, and dishwasher safe.
- Made of premium quality polypropylene plastic
- Superior chemical durability and heat tolerance
- Excellent optical clarity
- Raised 5 ml graduations and specifications
- Tapered spout for easy pouring
Polypropylene plastic beakers offer excellent chemical durability. They are microwavable, autoclavable, and dishwasher safe for easy cleaning and sterilization; can withstand temperatures between −20 °C to 135 °C. Beakers have raised, five ml graduation lines. Pack of 12.
Autoclavable, microwavable, and dishwasher safe.
- Made of premium quality polypropylene plastic
- Superior chemical durability and heat tolerance
- Excellent optical clarity
- Raised 5 ml graduations and specifications
- Tapered spout for easy pouring
Polypropylene plastic beakers offer excellent chemical durability. They are microwavable, autoclavable, and dishwasher safe for easy cleaning and sterilization; can withstand temperatures between −20 °C to 135 °C. Beakers have raised, five ml graduation lines. Pack of 12.
Product Family Options
- CapacitéDésignationHauteurØ ext.Intervalle de graduation
- 50 ml12/pack61 mm (2.4")44 mm (1.7")5 ml-: -
- : CH0137AF
- 100 ml12/pack74 mm (2.9")56 mm (2.2")5 ml-: CH0137BF
- : CH0137BF
- 250 ml6/pack100 mm (3.9")72 mm (2.8")10 ml-: -
- 250 ml12/pack100 mm (3.9")72 mm (2.8")10 ml-: -
- : CH0137CPK6
- : CH0137CF
- 500 ml12/pack122 mm (4.8")92 mm (3.6")10 ml-: -
- : CH0137DF
- 1000 ml12/pack148 mm (5.8")117 mm (4.6")20 ml-: -
- 1000 ml12/pack148 mm (5.8")117 mm (4.6")20 mlPack of 12$116.20: CH0137EF
- 2000 ml12/pack186 mm (7.3")148 mm (5.8")50 ml-: -
- 2000 ml12/pack186 mm (7.3")148 mm (5.8")50 mlPack of 12$136.10: CH0137FF
- : CH0137G