Eisco LDPE Dropping Bottles, Pack of 12
Fournisseur: Wards
Certificats
À propos de cet article
Dispenses precise uniform drops of liquid.
- Made of translucent low density polyethylene plastic (LDPE)
- Screw cap with a built in dropper nozzle allows for small, uniform drops of liquid
- Attached cover for nozzle prevents spills
Nozzle cover teathered to cap prevents leakage and spills.
Packaging: Comes in packs of 12 bottles.
Frequently Bought Together