Checker Plus® pH Tester, Electron Microscopy Sciences
Checker Plus® pH Tester, Electron Microscopy Sciences
Certificates
Checker Plus® pH Tester, Electron Microscopy Sciences
Checker Plus® pH Tester, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Checker Plus electronic pH tester supplies you with fast and accurate readings on a large LCD. The electrode can be replaced, allowing the tester to last for years.
  • Large and easy-to-read LCD
  • High accuracy with 0.01 pH resolution
  • Automatic, one or two points calibration
  • Battery life: 1,000 hours of continuous use
Modern Substitute for Litmus Paper with Replaceable Electrode. The extended family of Checker® pH testers now includes standard gel-filled electrodes with built-in BNC or screw type connectors.
Checker Plus is great for environmental, field and laboratory testing. Also suitable for checking electrodes in pipes and tanks and various laboratory applications.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Discontinued
    Supplier Number: -
  • Checker Plus complete set with HI1271 electrode
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $289.66
    $289.66
    Each
       $289.66
    Supplier Number: 70699-02
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Connector
    Availability
    Price
  • HI 1271 replacement electrode
    HI 1271 replacement electrode
    100493-012
    Model 70699-02
    Screw type
    Log in pour la disponibilité
    Each - $57.34