Klercide™ triple bucket system
Klercide™ triple bucket system
Catalog # 76554-226
Supplier: ECOLAB LIFE SCIENCES
Supplier #: 10056357
Jump to:
Product Details & Documents
Ecolab’s Klercide™ Triple Bucket System features a unique controlled application wringer which enables the even distribution of disinfectant over cleanroom surfaces. This enables accurate, repeatable results which help minimize operator error and enhance compliance with cGMP.
- Includes “click wringer” for controlled application that helps prevent potential fluid application variations between operators and ensures the optimum cleaning and disinfection results every time
- Includes 3 large buckets for segregation of solutions and waste, 4 small buckets for the top of the trolley and Klercide stainless steel flat mop frame and handle
- Fully autoclavable
Specifications
- Désignation:Klercide™ triple bucket system