Klercide™ mop wipe frame, stainless steel
Catalog # 76554-276
Supplier: ECOLAB LIFE SCIENCES
Specifications
- Description:Mop wipe frame, incl. one head and one handle
- Material:Stainless steel
- Cat. no.:76554-276
Specifications
About this item
This stainless steel mop frame features a stiff movable flat top.
- Fully autoclavable
- Made from stainless steel with stiff, moveable flat mop head frame
- Designed for use with Klercide™ pre-saturated mop wipes
- Supplied Gamma irradiated
Option of adding Klercide™ silicone pad for better contact on uneven surfaces.