Klercide™ Mop Frame and Handle, Stainless Steel
Catalogue # 76554-274
Fournisseur: ECOLAB LIFE SCIENCES
Spécifications
- Description:Mop frame and handle, extendable, incl. one head and one handle
- Length:1,0 - 1,5 m
- Material:Stainless steel
- Cat. no.:76554-274
Spécifications
À propos de cet article
This stainless steel mop frame and handle features a stiff movable flat top.
- Fully autoclavable
- Made from stainless steel with stiff, moveable flat mop head frame
- Designed for use with Klercide™ pre-saturated mop wipes
- Supplied Gamma irradiated
The extending handle makes it easy to reach high cleanroom ceilings and can be adjusted from 1.0 to 1.5 m long for ease of use.
Option of adding Klercide™ silicone pad for better contact on uneven surfaces.