Klercide™ Isolator Cleaning Tool
Isolator cleaning tool, incl. one head and two handles
Catalog # 76554-220
Supplier: ECOLAB LIFE SCIENCES
Fournisseur #: 92023002
Jump to:
Product Details & Documents
The Klercide™ Isolator Cleaning Tool and Cleaning Pads are specifically designed for isolator, laminar flow cabinet cleaning and other cleanroom areas where access is restricted.
- Fully autoclavable
- Lightweight for ease of use
- Made from 316 stainless steel with 600 mm reach
- Supplied cleanroom processed, double bagged and sterilized by gamma irradiation
- Compatible with the Klercide™ range of alcohols and biocides
The Klercide™ Isolator and Laminar Flow Cabinet Cleaning Tool consists of a two-part 316 stainless steel handle and a specially shaped head.
Designed for use with sterile single use cleaning pads.
Spécifications
- Désignation:Isolator cleaning tool, incl. one head and two handles