COSA™ PUR 80 surfactant additive
76554-212
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Size:5 gallon
- Cat. no.:76554-212
- Packaging:Pail
COSA™ PUR 80 is a liquid surfactant additive.
- Used as a CIP additive to enhance the cleaning effect of alkaline and acid cleaners for stubborn soils.
- Excellent removal of organic soiling such as fats present in residues of creams, ointments, emulsions, oils, and tablet coatings in CIP and manual applications.
- Supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation.
COSA™ PUR 80 is a neutral cleaning booster based on surfactants formulated for the removal of high fat-containing and water free-formulations in pharmaceutical processing equipment.