COSA™ CIP 94 Alkaline Cleaner
Fournisseur: ECOLAB LIFE SCIENCES
Certificats
À propos de cet article
COSA™ CIP 94 is a unique alkaline cleaner.
- Enhances Safety and Effectiveness of Cleaning
- Solubilizes and suspends organic soils, such as oil-in-water and water-in-oil emulsions, fatty residues and oils.
- Removes water and oil-soluble components of many cosmetic and pharmaceutical formulations.
- Eliminates the need for solvents during the cleaning process.
- Supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation.
COSA™ CIP 94 is a unique alkaline cleaner, formulated to remove difficult soils commonly found in the cosmetic and pharmaceutical industries. Examples of these include foundation, lipstick, sunscreen, and pharmaceutical ointments.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together