COSA™ CIP 72 liquid acid cleaner
Supplier: ECOLAB LIFE SCIENCES
Certificates
About this item
COSA™ CIP 72 is a liquid acid cleaner.
- Excellent removal of inorganic soiling.
- Free of phosphates, nitrogen, and surfactants.
- Aids rinsing of surfactants and alkalinity agents when used after an alkaline detergent wash.
- Supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation.
Ecolab COSA™ CIP 72 is an acid cleaner based on organic acids - free of phosphates, nitrogen, and surfactants for the removal of minerals and acid-soluble residues. Applicable for clean in place (CIP) pharmaceutical processing equipment including piping mixers, fluid beds, tanks, and storage vessels.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together