Quantum Levitation Extended Duration Levitators
À propos de cet article
The quantum enhanced-duration levitators are the perfect tool for science demonstrations and visual display. Engineered with years of experience and knowhow in cryogenics, these levitators offer the optimal balance between levitation strength and levitation time.
- Quantum extended duration levitator
- Plastic tweezers
- User manual
These new levitators offer 2 to 3× times longer levitation times compared to our standard ones. This means you’ll have more time to explain and discuss the physics or just appreciate their magic instead of constantly refilling with liquid nitrogen.
2 Options Available Below
Product Details & Documents
The quantum enhanced-duration levitators are the perfect tool for science demonstrations and visual display. Engineered with years of experience and knowhow in cryogenics, these levitators offer the optimal balance between levitation strength and levitation time.
- Quantum extended duration levitator
- Plastic tweezers
- User manual
These new levitators offer 2 to 3× times longer levitation times compared to our standard ones. This means you’ll have more time to explain and discuss the physics or just appreciate their magic instead of constantly refilling with liquid nitrogen.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Quantum extended duration levitator, medEach$1,646.29Pack of 1$1,454.07Supplier Number: 15-2
- Superconductor large extended durationEach$2,575.09Pack of 1$2,273.84Supplier Number: 15-3