Evergreen Tissue Processing/Embedding Cassettes, Caplugs Evergreen
À propos de cet article
Caplugs Evergreen tissue processing/embedding cassettes are used in the processing, embedding, sectioning, and long term storage of tissue and biopsy specimens.
- High density acetal construction
- Acetal is resistant to histological solvents such as alcohol and xylene
- Acetal will also keep the specimen submerged
- Modified ball-and-socket hinge design for ease of opening and for detachment
- Cassettes are available unassembled and bulk packed
Tissue processing/embedding cassettes feature efficient flow-through holes (1×1 mm) to maximize fluid exchange and ensure proper drainage.
2 Options Available Below
Product Details & Documents
Caplugs Evergreen tissue processing/embedding cassettes are used in the processing, embedding, sectioning, and long term storage of tissue and biopsy specimens.
- High density acetal construction
- Acetal is resistant to histological solvents such as alcohol and xylene
- Acetal will also keep the specimen submerged
- Modified ball-and-socket hinge design for ease of opening and for detachment
- Cassettes are available unassembled and bulk packed
Tissue processing/embedding cassettes feature efficient flow-through holes (1×1 mm) to maximize fluid exchange and ensure proper drainage.
Product Family Options
Informations sur le produit
- DésignationMatièreColorAvailabilityPrice
- 4-Compartment biopsy cassette (1 mm holes)AcetalGreenLog in pour la disponibilitéBag of 500$528.79Supplier Number: 258-4248-G90
- Supplier Number: 258-4249-H90