À propos de cet article
The Rheo Continuous Liner is an excellent pack-off solution for transferring materials from a work process into a transport container while maintaining a high level of containment. Continuous Liners come in various sizes and can be configured to work with existing process equipment.
- Constructed from RheoFlex 302 polyethylene material
- FDA approved for packaging
- USP and EP compliant
- 21 CFR 177.1520 compliant
- Matte finish
- Anti-static film
Continuous Liners keep the powder pack-off process clean and reduce the risk of cross contamination within the process suite. This technology can also provide a safe bag-in/bag-out option for transferring items into and out of an isolator.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.