Masterflex® I/P® MasterSense® Pump Systems
Masterflex® I/P® MasterSense® Pump Systems
Masterflex® I/P® MasterSense® Pump Systems
Certificates
Masterflex® I/P® MasterSense® Pump Systems
2 Options Available Below
Product Details & Documents
Versatile pumps for process applications – with an intuitive touch-screen interface and advanced connectivity.
  • Powerful, space-efficient drives will accept all I/P pump head and tubing options
  • 17.8 cm (7") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
  • Options for connectivity and monitoring include EtherNet/IP™, Profibus, analog signal or MasterflexLive,
  • Three-level user access – gives you the option of controlling who can operate the pump and at what level.
  • Continuous run, volume dispense, and time dispense modes – batch control and interval/off time in Volume and Time modes
  • This drive offers a broad flow range, from 0.0006 to 19 LPM, adaptable to a variety of applications and workflows – stack up to two pump heads for expanded flow
  • Drive housing is constructed of 316 stainless steel – IP66 and NEMA 4X rated for washdown
  • Real-time pressure sensing and alarms increase process reliability and frees up operator time
These Masterflex® I/P® MasterSense™ drives feature high precision, repeatable flow control, and reliable performance, you can have it all in an efficient footprint. Open-head sensor technology stops the drive motor when pump head is opened, or connect it to a remote leak detector to stop the pump anytime a leak is detected in the fluid path.
Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000:1 turndown ratio. Bidirectional flow and self-priming capabilities allow for smooth, seamless operation.
Pressure sensor integration feature brings single point pressure values to the pump interface in real time. Provide extra reliability to your process by configuring high/low set point alarms. Alarm can be set to trigger either a warning and/or pump shut off. Order required accessories for simple plug and play functionality: choose the MasterSense MP1 adaptor kit for single-use applications, the MasterSense MP30 reusable pressure transducers for repeat use.
MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps; get real-time notifications of pump parameters – including speed, flow rate, dispense volume, and more. You can monitor critical processes running 24/7 without the need for onsite personnel while notifications alert you to operating conditions and error messages. Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive.
Certifications: UL, cUL, CE.
Ordering information: Each model includes a country specific plug/cord set; choose the catalog number with desired country plug type. All units include an IEC 320 socket with a detachable standard US three prong power cable. See country plug description https://us.vwr.com/cms/tech-article-ac-power-plugs-sockets-universal-pattern-guide.
Delivery information: Includes drive, pump head, tubing, and 1.8 m (6 ft.) line cord with IEC 320/CEE22 socket. These models are shipped with country specific plug/cord set; please order the specific catalog number with the desired plug type.
Specifications for Product Family
Specifications
  • Model:
  • 77966-00
  • 77966-00
  • 77966-10
  • 77966-10
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • No. of rollers:
  • 3
  • 3
  • 3
  • 3
  • Control type:
  • Digital
  • Digital
  • Digital
  • Digital
  • Min. flow rate per channel:
  • 0.001 L/min
  • 0.001 L/min
  • 0.001 L/min
  • 0.001 L/min
  • Max. flow rate per channel:
  • 8000 ml/min
  • 8 L/min
  • 8000 ml/min
  • 8 L/min
  • Min. flow rate:
  • 0.001 L/min
  • 0.001 L/min
  • 0.001 L/min
  • 0.001 L/min
  • Drive IP rating:
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • Anti-drip:
  • Yes
  • Yes
  • Yes
  • Yes
  • Cloud connected:
  • Yes
  • Yes
  • Yes
  • Yes
  • Digital dispensing:
  • Yes
  • Yes
  • Yes
  • Yes
  • Drive:
  • 77421-00 I/P® MasterSense™ drive with EtherNet/IP™ and MasterflexLive®
  • 77421-00 I/P® MasterSense™ drive with EtherNet/IP™ and MasterflexLive®
  • 77421-02 I/P® MasterSense™ drive with Profibus and MasterflexLive®
  • 77421-02 I/P® MasterSense™ drive with Profibus and MasterflexLive®
  • Min. RPM:
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • Open-head sensor:
  • Yes
  • Yes
  • Yes
  • Yes
  • Pump series:
  • I/P
  • I/P
  • I/P
  • I/P
  • Remote I/O:
  • EtherNet/IP, MasterflexLive, Serial (ASCII), 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • EtherNet/IP, MasterflexLive, Serial (ASCII), 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • Profibus, MasterflexLive, Serial (ASCII), 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • Profibus, MasterflexLive, Serial (ASCII), 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • Sensor input:
  • Pressure, 0 to 20 mA, 0 to 10 V
  • Pressure, 0 to 20 mA, 0 to 10 V
  • Pressure, 0 to 20 mA, 0 to 10 V
  • Pressure, 0 to 20 mA, 0 to 10 V
  • Sensor input accuracy (aux port):
  • ±0.2% full scale
  • ±0.2% full scale
  • ±0.2% full scale
  • ±0.2% full scale
  • Reversible motor:
  • Yes
  • Yes
  • Yes
  • Yes
  • Tubing:
  • 06434-73 C-Flex ULTRA® I/P 73 tubing 3 m (10')
  • 06434-73 C-Flex ULTRA® I/P 73 tubing 3 m (10')
  • 06434-73 C-Flex ULTRA® I/P 73 tubing 3 m (10')
  • 06434-73 C-Flex ULTRA® I/P 73 tubing 3 m (10')
  • Tubing sizes accepted:
  • I/P® 26, 73, 82
  • I/P® 26, 73, 82
  • I/P® 26, 73, 82
  • I/P® 26, 73, 82
  • Washdown capable:
  • Yes
  • Yes
  • Yes
  • Yes
  • Frequency:
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • Voltage:
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Specifications
Product Family Options
US plug models
Product Information
  • Désignation
    Plug Type
    Débit maximal
    Hauteur
    Longueur
    Width
    Pump Head
    Puissance
    Max. RPM
    Matériau du boîtier
    Availability
    Price
  • MFLX77966-00
    Masterflex® I/P® MasterSense™ Process Pump, EtherNet/IP™ compatible with MasterflexLive®, with I/P® Easy-Load® pump head for precision tubing
    US
    8000 ml/min
    346 mm (13.6")
    432 mm (17")
    260 mm (10.2")
    77602-30 I/P® Easy-Load® head with open-head sensor
    115/230 VAC
    650 rpm
    316 stainless steel -- IP66 and NEMA 4X rated for washdown
    Log in for availability
    Log in for availability
    Each
    Each - $17,550.00
    $17,550.00
    Each
       $17,550.00
    Supplier Number: 77966-00
    Specifications:
    MasterflexLive®
    2 year warranty
    cUL Listed
    CE Compliant
    UL Listed
    More
    Specifications
    Cat. No.
    MFLX77966-00
    Model
    77966-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Digital
    Min. flow rate per channel
    0.001 L/min
    Max. flow rate per channel
    8000 ml/min
    Min. flow rate
    0.001 L/min
    Drive IP rating
    IP 66
    Anti-drip
    Yes
    Cloud connected
    Yes
    Digital dispensing
    Yes
    Drive
    77421-00 I/P® MasterSense™ drive with EtherNet/IP™ and MasterflexLive®
    Min. RPM
    0.1 rpm
    Open-head sensor
    Yes
    Pump series
    I/P
    Remote I/O
    EtherNet/IP, MasterflexLive, Serial (ASCII), 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Sensor input
    Pressure, 0 to 20 mA, 0 to 10 V
    Sensor input accuracy (aux port)
    ±0.2% full scale
    Reversible motor
    Yes
    Tubing
    06434-73 C-Flex ULTRA® I/P 73 tubing 3 m (10')
    Tubing sizes accepted
    I/P® 26, 73, 82
    Washdown capable
    Yes
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    MasterflexLive®
    2 year warranty
    cUL Listed
    CE Compliant
    UL Listed
  • MFLX77966-00-USN
    Masterflex® I/P® MasterSense™ Process Pump, EtherNet/IP™ compatible with MasterflexLive®, with I/P® Easy-Load® pump head for precision tubing
    USN
    8 L/min
    346 mm (13.6")
    432 mm (17")
    260 mm (10.2")
    77602-30 I/P® Easy-Load® head with open-head sensor
    115/230 VAC
    650 rpm
    316 stainless steel -- IP66 and NEMA 4X rated for washdown
    Discontinued
    Supplier Number: -
    Specifications:
    MasterflexLive®
    2 year warranty
    cUL Listed
    CE Compliant
    Irregular Voltage
    UL Listed
    More
    Specifications
    Cat. No.
    MFLX77966-00-USN
    Model
    77966-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Digital
    Min. flow rate per channel
    0.001 L/min
    Max. flow rate per channel
    8 L/min
    Min. flow rate
    0.001 L/min
    Drive IP rating
    IP 66
    Anti-drip
    Yes
    Cloud connected
    Yes
    Digital dispensing
    Yes
    Drive
    77421-00 I/P® MasterSense™ drive with EtherNet/IP™ and MasterflexLive®
    Min. RPM
    0.1 rpm
    Open-head sensor
    Yes
    Pump series
    I/P
    Remote I/O
    EtherNet/IP, MasterflexLive, Serial (ASCII), 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Sensor input
    Pressure, 0 to 20 mA, 0 to 10 V
    Sensor input accuracy (aux port)
    ±0.2% full scale
    Reversible motor
    Yes
    Tubing
    06434-73 C-Flex ULTRA® I/P 73 tubing 3 m (10')
    Tubing sizes accepted
    I/P® 26, 73, 82
    Washdown capable
    Yes
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    MasterflexLive®
    2 year warranty
    cUL Listed
    CE Compliant
    Irregular Voltage
    UL Listed
  • MFLX77966-10
    Masterflex® I/P® MasterSense™ Process Pump, Profibus compatible with MasterflexLive®, with I/P® Easy-Load® pump head for precision tubing
    US
    8000 ml/min
    346 mm (13.6")
    432 mm (17")
    260 mm (10.2")
    77602-30 I/P® Easy-Load® head with open-head sensor
    115/230 VAC
    650 rpm
    316 stainless steel -- IP66 and NEMA 4X rated for washdown
    Log in for availability
    Log in for availability
    Each
    Each - $17,590.00
    $17,590.00
    Each
       $17,590.00
    Supplier Number: 77966-10
    Specifications:
    MasterflexLive®
    2 year warranty
    cUL Listed
    CE Compliant
    UL Listed
    More
    Specifications
    Cat. No.
    MFLX77966-10
    Model
    77966-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Digital
    Min. flow rate per channel
    0.001 L/min
    Max. flow rate per channel
    8000 ml/min
    Min. flow rate
    0.001 L/min
    Drive IP rating
    IP 66
    Anti-drip
    Yes
    Cloud connected
    Yes
    Digital dispensing
    Yes
    Drive
    77421-02 I/P® MasterSense™ drive with Profibus and MasterflexLive®
    Min. RPM
    0.1 rpm
    Open-head sensor
    Yes
    Pump series
    I/P
    Remote I/O
    Profibus, MasterflexLive, Serial (ASCII), 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Sensor input
    Pressure, 0 to 20 mA, 0 to 10 V
    Sensor input accuracy (aux port)
    ±0.2% full scale
    Reversible motor
    Yes
    Tubing
    06434-73 C-Flex ULTRA® I/P 73 tubing 3 m (10')
    Tubing sizes accepted
    I/P® 26, 73, 82
    Washdown capable
    Yes
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    MasterflexLive®
    2 year warranty
    cUL Listed
    CE Compliant
    UL Listed
  • MFLX77966-10-USN
    Masterflex® I/P® MasterSense™ Process Pump, Profibus compatible with MasterflexLive®, with I/P® Easy-Load® pump head for precision tubing
    USN
    8 L/min
    346 mm (13.6")
    432 mm (17")
    260 mm (10.2")
    77602-30 I/P® Easy-Load® head with open-head sensor
    115/230 VAC
    650 rpm
    316 stainless steel -- IP66 and NEMA 4X rated for washdown
    Discontinued
    Supplier Number: -
    Specifications:
    MasterflexLive®
    2 year warranty
    cUL Listed
    CE Compliant
    Irregular Voltage
    UL Listed
    More
    Specifications
    Cat. No.
    MFLX77966-10-USN
    Model
    77966-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Digital
    Min. flow rate per channel
    0.001 L/min
    Max. flow rate per channel
    8 L/min
    Min. flow rate
    0.001 L/min
    Drive IP rating
    IP 66
    Anti-drip
    Yes
    Cloud connected
    Yes
    Digital dispensing
    Yes
    Drive
    77421-02 I/P® MasterSense™ drive with Profibus and MasterflexLive®
    Min. RPM
    0.1 rpm
    Open-head sensor
    Yes
    Pump series
    I/P
    Remote I/O
    Profibus, MasterflexLive, Serial (ASCII), 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Sensor input
    Pressure, 0 to 20 mA, 0 to 10 V
    Sensor input accuracy (aux port)
    ±0.2% full scale
    Reversible motor
    Yes
    Tubing
    06434-73 C-Flex ULTRA® I/P 73 tubing 3 m (10')
    Tubing sizes accepted
    I/P® 26, 73, 82
    Washdown capable
    Yes
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    MasterflexLive®
    2 year warranty
    cUL Listed
    CE Compliant
    Irregular Voltage
    UL Listed
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Cable assembly for remote control, 31-pin round connector and 7.9 m (25') cable with stripped wire ends
    Cable assembly for remote control, 31-pin round connector and 7.9 m (25') cable with stripped wire ends
    MFLX07575-80
    7.3 m (24 ft.)
    Log in for availability
    Each (1Feet) - $376.00
Product Information
  • Catalog #
    Availability
    Price
  • Footswitch, on/off operation, wash-down; 31-pin round, 6' cable
    Footswitch, on/off operation, wash-down; 31-pin round, 6' cable
    MFLX07575-84
    Log in for availability
    Each (6Feet) - $1,050.00
Product Information
  • Catalog #
    Availability
    Price
  • Serial coms cable USB-A - USB-A, 8'
    Serial coms cable USB-A - USB-A, 8'
    MFLX78006-90
    Log in for availability
    Each - $157.00
Product Information
  • Catalog #
    Includes
    Availability
    Price
  • Masterflex® MasterSense® MP30 Pressure Sensor with 1¹/₂" Sanitary Tri-Clamp Process Connection, for MasterSense® Pump Systems
    Masterflex® MasterSense® MP30 Pressure Sensor with 1¹/₂" Sanitary Tri-Clamp Process Connection, for MasterSense® Pump Systems
    MFLX07526-82
    6' cable
    Log in for availability
    Each - $2,100.00
Product Information
  • Catalog #
    Includes
    Availability
    Price
  • Sensor kit; Masterflex® MP1 PendoTECH™ single-use pressure sensors kit for MasterSense® pump systems
    Sensor kit; Masterflex® MP1 PendoTECH™ single-use pressure sensors kit for MasterSense® pump systems
    MFLX07526-91
    7526-90 MP1 adaptor, ¹/₄ and ³/₈" Luer to hose barb tee fittings
    Log in for availability
    Each - $2,100.00
Product Information
  • Catalog #
    Load Capacity
    Availability
    Price
  • Masterflex® bioprocess pump cart
    Masterflex® bioprocess pump cart
    MFLX77112-90
    550 kg (1200 lbs.)
    Log in for availability
    Each - $6,540.00