VWR® Standard Laboratory Chest Freezer with Manual Defrost (15 cu. ft.)
Catalog # 76580-202
Supplier: VWR International
Specifications
- Temperature Range:‒15...‒25 °C
- Capacity:424 L (15 cu.ft.)
- Description:Standard series laboratory chest freezer manual defrost, 15 CF
- Cat. No.:76580-202
- Electrical:115 V, 60 Hz, 5 A, ¼ HP
- Defrost:Manual
- Door type:1 solid door, right hinged
- Shelves/drawers:2 Removable baskets
- Weight:89 kg (197 lbs.)
- Exterior W×D×H:1422×749×892 mm (56×29½×35⅛")
- Interior W×D×H:1254×546×651 mm (49⅜×21½×25⅝")
Specifications
About this item
These VWR® Standard Series manual defrost chest freezers provide quality cold storage at an economical pricepoint.
- Mechanical thermostat
- Keyed door lock
- High density foam insulation
- Two removable baskets
- Manual defrost
These manual defrost laboratory chest freezers offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, and precise temperature monitoring through microprocessor control. The Standard line provides multi-functional features in a cost-effective design. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Material: White powder coat
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.