VWR® Plus Series Compact Laboratory Refrigerators with Natural Refrigerants
Supplier: VWR International
Certificates
About this item
These VWR® Plus Series compact laboratory refrigerators offer premium cold storage with superior temperature control.
- Microprocessor temperature controller
- Digital temperature display
- Audible and visual alarm systems
- LED interior lighting, and adjustable shelving
- Keyed door lock
- Natural, hydrocarbon refrigerant
- ENERGY STAR certified*
These compact glass door laboratory refrigerators offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, probe access, and precise temperature monitoring through microprocessor control. The Plus Series line includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Material: White powder coat
Certifications: UL/C-UL listed.
*Select models ENERGY STAR certified: 76580-148, 76580-150
*Select models ENERGY STAR certified: 76580-148, 76580-150
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Specifications
- Catalog No:
- 76580-150
- 76580-148
- Environmentally Preferable:
- Energy Efficient
- Energy Efficient
- Cat. No.:
- 76580-150
- 76580-148
- Defrost:
- Cycle
- Cycle
- Shelves/drawers:
- 6 adjustable shelves
- 6 adjustable shelves
- Door type:
- 1 solid door, right hinged
- 1 glass door, right hinged
- Electrical:
- 115 V, 60 Hz, ⅓ HP, 2.3 A
- 115 V, 60 Hz, ⅓ HP, 2.3 A
- Weight:
- 84 kg (183 lbs.)
- 83 kg (183 lbs.)
- Environmental certification:
- Uses Natural Hydrocarbon refrigerants
- Uses Natural Hydrocarbon refrigerants