Michels Solution , Statlab | Avantor
Michels Solution , Statlab
Supplier:   StatLab
Michels Solution , Statlab
Supplier:  StatLab
Certificates
Michels Solution , Statlab
Michels Solution , Statlab
3 Options Available Below
Product Details & Documents
Michels Solution is used to preserve and transport specimens for examination in immunofluorescence studies.
  • While not a fixative, specimens can be held at room temperature for five days in transport medium before processing
  • This simple salt solution maintains pH, but does not kill most pathogens
  • Specimens received in transport medium should be washed in three changes of Michels Wash Solution (10 minutes for each wash)
  • Not suitable for FISH studies
Part of the StatLab line of formalin alternatives.
Manufactured in the USA
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Michels Solution
    20 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 96 (20ml)
    Case of 96 (20ml) - $366.27
    $366.27
    Log in pour la disponibilité
    Each(20ml)
    Each(20ml) - $366.27
    $366.27
    Case of 96 (20ml)
       $366.27
    Each(20ml)
       $366.27
    Supplier Number: MS0507
  • Michels Solution
    7 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100 (7ml)
    Case of 100 (7ml) - $154.99
    $154.99
    Log in pour la disponibilité
    Each(7ml)
    Each(7ml) - $154.99
    $154.99
    Case of 100 (7ml)
       $154.99
    Each(7ml)
       $154.99
    Supplier Number: MS0735
  • Michels Solution
    32 oz
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (32oz)
    Each (32oz) - $115.01
    $115.01
    Each (32oz)
       $115.01
    Supplier Number: MS-32