VWR® Plus Series Solid Door Laboratory Refrigerators
À propos de cet article
These VWR® Plus Series laboratory refrigerators offer premium cold storage solutions with superior temperature control.
- Microprocessor temperature controller
- Digital temperature display and alarm module
- Audible, visual, senor error, and power failure alarms
- Keyed door lock, LED interior lighting, and adjustable shelving
- Alarm validation and alarm mute with ring back
- Natural, hydrocarbon refrigerant
- ENERGY STAR certified*
These VWR® refrigerators are designed with premium features for laboratory use. They feature a series of alarms for product protection and remote alarm contacts. Units employ forced draft air circulation for superior temperature control. These refrigerators utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Material: White powder coat
Certifications: ETL Listed.
*Select models ENERGY STAR certified: 76579-106, 76579-110, 76579-114, 76579-122
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
7 Options Available Below
Product Details & Documents
These VWR® Plus Series laboratory refrigerators offer premium cold storage solutions with superior temperature control.
- Microprocessor temperature controller
- Digital temperature display and alarm module
- Audible, visual, senor error, and power failure alarms
- Keyed door lock, LED interior lighting, and adjustable shelving
- Alarm validation and alarm mute with ring back
- Natural, hydrocarbon refrigerant
- ENERGY STAR certified*
These VWR® refrigerators are designed with premium features for laboratory use. They feature a series of alarms for product protection and remote alarm contacts. Units employ forced draft air circulation for superior temperature control. These refrigerators utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Material: White powder coat
Certifications: ETL Listed.
*Select models ENERGY STAR certified: 76579-106, 76579-110, 76579-114, 76579-122
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Specifications for Product Family
Spécifications
- Catalog No:
- 76579-102
- 76579-106
- 76579-110
- 76579-114
- 76579-118
- 76579-122
- 76579-126
- Environmentally Preferable:
- Safer Human and Environmental Health|Energy Efficient
- Safer Human and Environmental Health|Energy Efficient
- Safer Human and Environmental Health|Energy Efficient
- Safer Human and Environmental Health|Energy Efficient
- Safer Human and Environmental Health|Energy Efficient
- Safer Human and Environmental Health|Energy Efficient
- Safer Human and Environmental Health|Energy Efficient
- Defrost:
- Cycle
- Cycle
- Cycle
- Cycle
- Cycle
- Cycle
- Cycle
- Shelves/drawers:
- 3 adjustable shelves (adjustable in ½" increments)
- 3 adjustable shelves (adjustable in ½" increments)
- 4 adjustable shelves (adjustable in ½" increments)
- 4 adjustable shelves (adjustable in ½" increments)
- 8 adjustable shelves (adjustable in ½" increments)
- 8 adjustable shelves (adjustable in ½" increments)
- 12 adjustable shelves (adjustable in ½" increments)
- Door type:
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- 2 solid doors, right hinged
- 2 solid doors, right hinged
- 2 solid doors, right hinged
- Electrical:
- 115 V, 60 Hz, 3 A, ⅕ HP
- 115 V, 60 Hz, 3 A, ⅕ HP
- 115 V, 60 Hz, 3 A, ⅕ HP
- 115 V, 60 Hz, 3 A, ⅕ HP
- 115 V, 60 Hz, 4.5 A, ¼ HP
- 115 V, 60 Hz, 4.5 A, ¼ HP
- 115 V, 60 Hz, 7.5 A, ½ HP
- Weight:
- 102 kg (227 lbs.)
- 126 kg (277 lbs.)
- 113 kg (250 lbs.)
- 156 kg (345 lbs.)
- 163 kg (360 lbs.)
- 202 kg (446 lbs.)
- 272 kg (600 lbs.)
- Environmental certification:
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
Spécifications
Product Family Options
Informations sur le produit
- DésignationCapacitéPlage de températureExterior W×D×HInterior W×D×HAvailabilityPrice
- Laboratory refrigerator upright solid door Plus series 12 CF340 L (12 cu.ft.)1...10 °C635×756×1619 mm (25×29¾×65¾")502×489×921 mm (19¾×19¼×36¼")Log in pour la disponibilitéEach$11,590.95Spécifications:
Safer Human and Environmental Health|Energy Efficient More
Specifications
Cat. No.76579-102Environmentally PreferableSafer Human and Environmental Health|Energy EfficientDefrostCycleShelves/drawers3 adjustable shelves (adjustable in ½" increments)Door type1 solid door, right hingedElectrical115 V, 60 Hz, 3 A, ⅕ HPWeight102 kg (227 lbs.)Environmental certificationUnits use natural, hydrocarbon refrigerantsSafer Human and Environmental Health|Energy Efficient - Laboratory refrigerator upright solid door Plus series 16 CF453 L (16 cu.ft.)1...10 °C635×756×2007 mm (25×29¾×79")508×495×1251 mm (20×19½×49¼")Log in pour la disponibilitéEach$13,712.77Spécifications:
Safer Human and Environmental Health|Energy Efficient More
Specifications
Cat. No.76579-106Environmentally PreferableSafer Human and Environmental Health|Energy EfficientDefrostCycleShelves/drawers3 adjustable shelves (adjustable in ½" increments)Door type1 solid door, right hingedElectrical115 V, 60 Hz, 3 A, ⅕ HPWeight126 kg (277 lbs.)Environmental certificationUnits use natural, hydrocarbon refrigerantsSafer Human and Environmental Health|Energy Efficient - Laboratory refrigerator upright solid door Plus series 23 CF651 L (23 cu.ft.)1...10 °C683×883×2076 mm (26⅞×34¾×81¾")559×641×1334 mm (22×25¼×52½")Log in pour la disponibilitéEach$15,100.26Spécifications:
Safer Human and Environmental Health|Energy Efficient More
Specifications
Cat. No.76579-110Environmentally PreferableSafer Human and Environmental Health|Energy EfficientDefrostCycleShelves/drawers4 adjustable shelves (adjustable in ½" increments)Door type1 solid door, right hingedElectrical115 V, 60 Hz, 3 A, ⅕ HPWeight113 kg (250 lbs.)Environmental certificationUnits use natural, hydrocarbon refrigerantsSafer Human and Environmental Health|Energy Efficient - Laboratory refrigerator upright solid door Plus series 26 CF736 L (26 cu.ft.)1...10 °C721×927×2076 mm (28⅜×36½×81¾")603×711×1334 mm (23¾×28×52½")Log in pour la disponibilitéEach$15,209.49Spécifications:
Safer Human and Environmental Health|Energy Efficient More
Specifications
Cat. No.76579-114Environmentally PreferableSafer Human and Environmental Health|Energy EfficientDefrostCycleShelves/drawers4 adjustable shelves (adjustable in ½" increments)Door type1 solid door, right hingedElectrical115 V, 60 Hz, 3 A, ⅕ HPWeight156 kg (345 lbs.)Environmental certificationUnits use natural, hydrocarbon refrigerantsSafer Human and Environmental Health|Energy Efficient - Laboratory refrigerator upright solid door Plus series 36 CF1019 L (36 cu.ft.)1...10 °C1007×883×2076 mm (39⅝×34¾×81¾")886×600×1340 mm (34⅞×23⅝×52¾")Log in pour la disponibilitéEach$18,926.39Spécifications:
Safer Human and Environmental Health|Energy Efficient More
Specifications
Cat. No.76579-118Environmentally PreferableSafer Human and Environmental Health|Energy EfficientDefrostCycleShelves/drawers8 adjustable shelves (adjustable in ½" increments)Door type2 solid doors, right hingedElectrical115 V, 60 Hz, 4.5 A, ¼ HPWeight163 kg (360 lbs.)Environmental certificationUnits use natural, hydrocarbon refrigerantsSafer Human and Environmental Health|Energy Efficient - Laboratory refrigerator upright solid door Plus series 49 CF1388 L (49 cu.ft.)1...10 °C1372×883×2076 mm (54×34¾×81¾")1245×626×1334 mm (49×24⅝×52½")Log in pour la disponibilitéEach$21,723.08Spécifications:
Safer Human and Environmental Health|Energy Efficient More
Specifications
Cat. No.76579-122Environmentally PreferableSafer Human and Environmental Health|Energy EfficientDefrostCycleShelves/drawers8 adjustable shelves (adjustable in ½" increments)Door type2 solid doors, right hingedElectrical115 V, 60 Hz, 4.5 A, ¼ HPWeight202 kg (446 lbs.)Environmental certificationUnits use natural, hydrocarbon refrigerantsSafer Human and Environmental Health|Energy Efficient - Laboratory refrigerator upright solid door Plus series 72 CF2039 L (72 cu.ft.)1...10 °C2057×864×2076 mm (81×34×81¾")1930×629×1327 mm (76×24¾×52¼")Log in pour la disponibilitéEach$30,458.49Spécifications:
Safer Human and Environmental Health|Energy Efficient More
Specifications
Cat. No.76579-126Environmentally PreferableSafer Human and Environmental Health|Energy EfficientDefrostCycleShelves/drawers12 adjustable shelves (adjustable in ½" increments)Door type2 solid doors, right hingedElectrical115 V, 60 Hz, 7.5 A, ½ HPWeight272 kg (600 lbs.)Environmental certificationUnits use natural, hydrocarbon refrigerantsSafer Human and Environmental Health|Energy Efficient