SureSTART™ Total Recovery Glass Screw Top 1.5 ml Microvials for <2 ml Samples, Level 3 High Performance Applications
Choose Thermo Scientific™ SureSTART™ 1.5 ml total recovery glass screw top microvials, performance Level 3, when you need to maximize the injection volume for <2 ml samples.
- Specification Certified to ensure their physical dimensions and autosampler compatibility
- Meets all requirements of US, EU, JPN Pharmacopeia
- Seal with 9 mm screw caps and septa
- 100% traceable through the manufacturing process
- All Thermo Scientific SureSTART vials are dimensionally verified
- Inspected by automated optical and gauge inspection to ensure critical dimensions and tolerances
- Uniform and defect-free
Available in clear and amber glass, this one-piece total recovery microvials has a very low residual volume due to a specially-shaped deep and narrow inner reservoir which collects the liquid in the base so that compatible autosamplers can collect all but 1 to2 µl of sample while having a total sample volume of 1.2 ml and a residual volume of 1 µl. The outer dimensions for these vials are 12×32 mm. These performance Level 3 vials are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyzes.
: Certificate of Conformance available by lot number upon request.
Caps
Frequently Bought Together