Containers, Burdick & Jackson™ NOWPak® | Avantor
Containers, Burdick & Jackson™ NOWPak®
10 L, HDPE
Catalog # CABJNP508
Supplier:   Burdick & Jackson
Fournisseur #:  NP508
Containers, Burdick & Jackson™ NOWPak®
Catalog # CABJNP508
Supplier:  Burdick & Jackson
Containers, Burdick & Jackson™ NOWPak®
10 L, HDPE
Price
Renseignez-vous sur le prix
Log in pour la disponibilité
Each Case of 2
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Empty containers made from HDPE.
  • With closure
  • Maintains solvent purity, increases lab safety and maximizes efficiency when using larger solvent volumes
Accessory/spare part for the NOWPak® system.
Volume: 10 L (2.6 gallons)
Delivery information: Sold as a pair (2 × 10 L).
Spécifications
  • Capacité:
    10 L
  • Matière:
    HDPE