Vented Balance Enclosures, Yamato
Fournisseur: Yamato Scientific
Certificats
À propos de cet article
Self-contained units ideal for use handling fine powders, chemicals and biological products.
- Includes top mounted HEPA/blower filtration set-up to circulate internal chamber atmosphere, through the HEPA filter and into the laboratory
- Smooth curved, or rounded ABSTM airfoils provide gentle, unobstructed air flow through rear, and side, baffles and through top mounted HEPA filter
- Constructed with optically clear ³/₈" jeweled acrylic plastic
- Front viewing sash with multiple air foils for higher containment and draft deflection
- Includes phenolic base with superior chemical resistance and provides higher analytical balance stabilization
- Low vibrations, black base allows for easy powder detection
- Air flow alarm system
The enclosures feature adjustable front draft protection, have a side blank plate for optional bag-in/bag-out port, and include a face velocity alarm.
Certifications: CE certification.
Frequently Bought Together