Universal Pipette Tips With Filter, Racked, Sterile, United Scientific Supplies
470349-754
- Volume:300 µl
- Cat. no.:470349-754
Racked and sterile universal pipette tips with filter.
- Manufactured from high grade polypropylene
- Filter is 100% pure porous polyethylene
- 99% Bacterial Aerosol Filtration Efficiency (BFE), ASTM F21012 Non-cytotoxic: ISO 10993-5 MRM, Minimum Essential Media Elution, GLP Compliant
- Non-hemolytic: Modified ASTM F 756-08 Hemolysis Direct Contact GLP with Human Blood
- DNase, RNase, Pyrogen Free
Protect against cross-contamination using these racked, sterile filter pipette tips. These universal tips are manufactured from polypropylene and the filters are manufactured from polyethylene with a hydrophobic design to prevent liquids and aerosols from entering the body of the micropipet. These tips are non-cytotoxic, non-hemolytic, and have 99.9% Bacterial Aerosol Filtration Efficiency (BFE). DNase, RNase and Pyrogen free. Packaged in racks of 96 with 10 racks in each.