ContactUsLinkComponent
Bluing Agent Reserve, Statlab
Catalog # 76548-224
Supplier:  StatLab
CAS Number:  
undefined
Bluing Agent Reserve, Statlab
Catalog # 76548-224
Supplier:  StatLab
Supplier Number:  SL203
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Bluing Agent Reserve
  • Size:
    1 gallon
  • Cat. no.:
    76548-224
Specifications
About this item
Converts the soluble red component of hematoxylin into an insoluble blue.
  • A buffered alkaline rinse with a mild pH (8.0)
  • Ensures optimal staining and proper cellular contrast
  • Bluing reagent is premixed and ready-to-use.
Part of the StatLab Reserve stain line.
Manufactured in USA.