RheoCrimps, Rheo Flexibles (Division of Rheo Engineering)
Supplier: RHEO ENGINEERING LLC
Certificates
About this item
The RheoCrimp is an easy to use cut and crimp separation technology. These crimps offer a wide opening range for capturing bulky liner material. The RheoCrimp kit includes the tools needed for this crimp and cut process.
- Wide-range crimp capability
- Dual-tie design
- Easy alignment cut-notch for cutting operation
- Grip-tight locking rib to ensure integrity
The wrap-around design of a tie-wrap allows for the operator to easily envelop a wide range of material, with little twisting and bunching.
Ordering information: Kit includes Cable -Tie tensioner, 50 RheoCrimps, 50 zip ties, roll of GMP tape, and cutting shears for cutting between Cable-Ties. Each RheoCrimp pack includes 50 RheoCrimps.
Packaging: All components packaged in tidy carrying case.
Frequently Bought Together