United Scientific™ Overflow Can And Catch Bucket Set
470339-984
- Description:Overflow can and bucket set
- Cat. no.:470339-984
Aluminum catch can for a variety of experiments.
- Aluminum
- For use in Archimedes' Principle, density, and specific gravity experiments
An aluminum overflow can and a catch bucket are packaged as a convenient set for use in Archimedes' Principle, density, and specific gravity experiments. The overflow can measures 4ID×5" high and has a capacity of approximately 800 ml. Its spout is 0.5ID×1.5" long. The catch bucket with detachable aluminum handle measures 4×3" high.