ContactUsLinkComponent
United Scientific™ Overflow Can And Catch Bucket Set
  470339-984
undefined
United Scientific™ Overflow Can And Catch Bucket Set
  470339-984
 :  OFLSET
  • Description:
    Overflow can and bucket set
  • Cat. no.:
    470339-984
Aluminum catch can for a variety of experiments.
  • Aluminum
  • For use in Archimedes' Principle, density, and specific gravity experiments
An aluminum overflow can and a catch bucket are packaged as a convenient set for use in Archimedes' Principle, density, and specific gravity experiments. The overflow can measures 4ID×5" high and has a capacity of approximately 800 ml. Its spout is 0.5ID×1.5" long. The catch bucket with detachable aluminum handle measures 4×3" high.