J.T.Baker®, PCR 8-Tube Strips and 8-Cap Strips for qPCR
Fournisseur: Ritter, part of Avantor
Certificats
À propos de cet article
PCR 8-tube strips and 8-cap strips are made of high quality PP resin. Clear caps are compatible with qPCR application.
- Certified free from DNase, RNase and human DNA
- White 8-tube strips
- Ideal for low volume PCR and qPCR applications
- Low profile tubes help to reduce condensation
- Validated on Biorad CFX device
PCR strips and caps are manufactured under clean room conditions according to DIN EN ISO 14644-1:2016-06.
Ordering information: Corresponding 8-cap strips, clear to be ordered separately.
Spécifications
- Catalog No:
- 43001-0033
- 43001-0034
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)