Insect Spreading Board | Avantor
Insect Spreading Board
Insect spreading board
Catalog # 470346-234
Supplier:   ARCO
Fournisseur #:  4120300
Insect Spreading Board
Catalog # 470346-234
Supplier:  ARCO
Insect Spreading Board
Insect Spreading Board
Insect spreading board
Price
$29.95
Log in pour la disponibilité
Each
Product Details & Documents
Prepare your insects for mounting.
  • Easily adjustable width
  • Cork coating
Made of wood covered with thick cork sheet. The groove from 0 to 12 mm is smoothly adjustable, die-pressed adjustment chrome plated brass fittings.
Spécifications
  • Désignation:
    Insect spreading board