Insect Spreading Board
Insect spreading board
Catalog # 470346-234
Supplier: ARCO
Fournisseur #: 4120300
Jump to:
Product Details & Documents
Prepare your insects for mounting.
- Easily adjustable width
- Cork coating
Made of wood covered with thick cork sheet. The groove from 0 to 12 mm is smoothly adjustable, die-pressed adjustment chrome plated brass fittings.
Spécifications
- Désignation:Insect spreading board