Agate Mixer Beads, Retsch
: Retsch
These beads are for use with MM 200 Mixers from Retsch.
- Composition is approximately 99.9% SiO₂
- Degree of hardness of 7 Mohs
- Approximate density is 2600 kg/m³
Frequently Bought Together
These beads are for use with MM 200 Mixers from Retsch.