Bead Ruptor Bulk Beads, Omni International
Supplier: OMNI INTERNATIONAL
Certificates
About this item
Bead beating is the most effective and verstile lysis method available
Bulk lysing matrix bead media. Certified Rnase- and Dnase-free to ensure high analyte integrity. Designed for use on all Omni International Bead Ruptor platforms.
Omni International offers users the ultimate flexibility with a range of bead beating lysing matrices to support efficient homogenization of a wide range of samples.
Frequently Bought Together