Vac Elut SPS 24 Manifold, Agilent Technologies
Vac Elut 24 SPS manifold processes up to 24 SPE cartridges at the same time.
- Closed operation prevents cross contamination
- Stainless steel tips deliver maximum extract purity
- Range of rack sizes covers most tube configurations
The Vac Elut SPS 24 manifold allows you to process up to 24 SPE cartridges simultaneously. Like all Vac Elut manifolds, the SPS 24 is made from durable, solvent-resistant materials and engineered to last. The glass sides allow easy viewing of the entire sample collection process.
Frequently Bought Together