Disintegration Basket Covers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Agilent offers disintegration basket covers for the 100 automated disintegration apparatus.
Frequently Bought Together
VWR® Inserts for Wide Opening Vials
Supplier: VWR International
VWR® Screw Caps for 4 ml Screw-Thread Vials
Supplier: VWR International
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Supplier: MicroSolv Technology Corp
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
Plastic Vials, Caps and Convenience Kits for LCMS and HPLC
Supplier: MicroSolv Technology Corp
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Supplier: VWR International
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
WHEATON® High Performance Crimping Tool and Jaw Sets, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
CAP ALUMINUM FOR 1ML VIAL 11MM PK1000
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Supplier: MicroSolv Technology Corp
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
High Recovery Vials and Inserts, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Inserts for Standard Opening Crimp-Top Vial, 11 mm, Thermo Scientific
Supplier: Thermo Scientific
WHEATON® MicroLiter Snap-Top Vials, 11 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
VWR® Crimp Caps for 20 mm Clear Crimp Seal Headspace Vials
Supplier: VWR International