PUREZERO* HM4 Pleated Face Mask with Earloops
Catalogue # 76558-120
Fournisseur: O&M HALYARD CANADA, INC CA
Spécifications
- Description:PUREZERO* HM4 Pleated Face Mask with Earloops
- Cat. no.:76558-120
Spécifications
À propos de cet article
The PUREZERO* HM4 Pleated Mask with Earloops is a three-layer mask with nylon earloops that provides a high particle, bacterial, and viral filtration efficiency.
- Three-layer mask
- Nylon earloops
- For industrial and cleanroom use
- Recommended for ISO Class 4 and above applications
Masks feature an exclusive SO SOFT* biocomponent technology inner lining for comfort.