PUREZERO* HM4 Pleated Face Mask with Ties
Catalogue # 76558-124
Fournisseur: O&M HALYARD CANADA, INC CA
Spécifications
- Description:PUREZERO* HM4 Pleated Face Mask with Ties
- Cat. no.:76558-124
Spécifications
À propos de cet article
The PUREZERO* HM4 Pleated Mask with Ties is a three-layer mask with long ties that provides high particle, bacterial, and viral filtration efficiency.
- Three-layer mask
- Long ties for ease of donning
- For industrial and cleanroom use
- Recommended for ISO Class 4 and above applications
Masks feature an exclusive SO SOFT* biocomponent technology inner lining for comfort.