Bond Elut SAX Plates, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
Bond Elut SAX is a strong anion-exchange sorbent well suited to compounds such as carboxylic acids.
- Retains compounds that elute from weak anion-exchange sorbents
- Selectivity can be user-modified for increased flexibility
- Minimal nonpolar interactions
Bond Elut SAX is a strong anion-exchange sorbent ideally suited for the extraction of compounds, such as carboxylic acids, which may not retain effectively on weak anion-exchange sorbents.
Frequently Bought Together