Well Plate Tubes for Bond Elut Plexa PAX Plates, Agilent Technologies
Catalogue # CAAG5982-0602
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Spécifications
- Volume:—
- Description:Bond Elut Plexa PAX tubes, VersaPlate, 30 mg
- Cat. no.:CAAG5982-0602
Spécifications
À propos de cet article
Bond Elut Plexa PAX nonpolar/anion-exchange SPE is suitable for polar and nonpolar acidic analytes.
- Mixed-mode nonpolar polymeric anion exchanger offers high level of analyte selectivity
- Exclusion of endogenous interferences offers superior cleanliness and minimizes ion suppression
- Simple, single method for ease-of-use reduces method development time
Bond Elut Plexa PAX lifts the performance standard in analyte cleanup and reproducibility for polar and nonpolar acidic analytes. Plexa PAX particles are functionalized using a proprietary process, allowing anion-exchange loadings to be controlled with high reproducibility. This gives more robust performance across your compound study or method.