Well Plate Tubes for Bond Elut NH2 Plates, Agilent Technologies
Catalogue # CAAG75505050
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Spécifications
- Volume:1.8 ml
- Description:VersaPlate well plate tubes, NEXUS, 30 mg, PE frit (20 µm)
- Cat. no.:CAAG75505050
Spécifications
À propos de cet article
Bond Elut NH2 is a weak anion exchanger and a good choice for retention of very strong anions.
- Streamlined sample preparation for lipid analysis
- Features innovative EMR-Lipid sorbent technology
- Time savings compared to traditional liquid-liquid extraction methods
- mproved reproducibility compared to traditional liquid-liquid extraction methods, reducing the number of samples required for statistical evaluation
- 2 ml well plate design is easily automatable for even more streamlined lipid analysis
Bond Elut NH2 is a weaker anion exchanger than sorbents such as SAX and is therefore a better choice for retention of very strong anions. Similar to Bond Elut Diol and SI sorbents, Bond Elut NH2 is excellent for the separation of structural isomers.