Well Plate Tubes for Bond Elut C18 OH Plates, Agilent Technologies
Catalog # CAAG7551601C
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Volume:1.8 ml
- Description:VersaPlate well plate tubes, C18 OH, 100 mg, PE frit (20 µm)
- Cat. no.:CAAG7551601C
Specifications
About this item
Bond Elut C18 OH enhances retention of basic compounds and promotes metabolite fractionation.
- Silanol activity permits metabolite fractionation
- Tight QC tolerances deliver batch-to-batch reproducibility
- 50 Å pore size expands usage for higher molecular weight compounds
Bond Elut C18 OH is a nonendcapped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 phase has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to endcapped C18. The 150 Å pore size makes it a good choice for peptide extraction and higher molecular weight compounds.