Dissolution Sinkers, Agilent Technologies | Avantor
Dissolution Sinkers, Agilent Technologies
Dissolution Sinkers, Agilent Technologies
Certificates
Dissolution Sinkers, Agilent Technologies
Dissolution Sinkers, Agilent Technologies
2 Options Available Below
Product Details & Documents
A variety of dissolution sinkers are available depending on your dosage form.
  • Capsule wire, weights, and sinker baskets are used to retain a dosage form at the bottom of the vessel for testing when the dosage form is buoyant
  • A variety of options are available
  • The flexible capsule wire meets USP requirements and has a 0.032" diameter
  • Sinker baskets are offered in a variety of mesh sizes and made of stainless steel with press-on caps, using O-rings to seal the basket
  • Japanese Pharmacopoeia (JP) sinker baskets are available as an alternative
  • Three-prong capsule weights fit traditional capsule sizes one through three, and are sold individually
Capsule wire, weights, and sinker baskets are used to retain a solid dosage form at the bottom of the vessel for testing when the dosage form is buoyant. Depending on your dosage form, a variety of options are available.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Capsule weight wire, 316 stainless steel, 0.032" Ø, 50' roll
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $359.00
    $359.00
    Each
       $359.00
    Supplier Number: 12-3000
  • Sinker basket, Japanese Pharmacopoeia (JP), stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $518.00
    $518.00
    Each
       $518.00
    Supplier Number: 12-3070