Dissolution Evaporation Covers, Agilent Technologies
Dissolution Evaporation Covers, Agilent Technologies
Dissolution Evaporation Covers, Agilent Technologies
Certificates
Dissolution Evaporation Covers, Agilent Technologies
Dissolution Evaporation Covers, Agilent Technologies
11 Options Available Below
Product Details & Documents
A variety of evaporation covers are available to fit your apparatus and vessels.
  • The evaporation covers automatically travel with the apparatus driveunit using either the alignment posts or DDM assemblies
  • The standard evaporation cover on the 708-DS is designed to allow less than 1% evaporative loss over a 24-hour period under normal operating conditions
  • Agilent also offers other low loss covers that can reduce evaporative loss even further
A variety of evaporation covers are available to fit your apparatus and vessels. Evaporation plugs are used to minimize evaporation by sealing unused ports in the evaporation covers.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • EaseAlign Evaporation cover, Basket, for 705-DS, VK7000,7010
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 224,00 $
    224,00 $
    Each
       224,00 $
    Supplier Number: 12-6305
  • EaseAlign Evaporation cover, DDM, for 705-DS, VK7000,7010
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 203,00 $
    203,00 $
    Each
       203,00 $
    Supplier Number: 12-6310
  • EaseAlign Evaporation cover, for 705-DS, VK7000,7010
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 161,00 $
    161,00 $
    Each
       161,00 $
    Supplier Number: 12-6300
  • EaseAlign Evaporation cover, for use with 100/200 ml vessel, for 705-DS, VK7000,7010
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 224,00 $
    224,00 $
    Each
       224,00 $
    Supplier Number: 12-6315
  • EaseAlign Evaporation cover, low actinic - red, for 705-DS, VK7000,7010
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 199,00 $
    199,00 $
    Each
       199,00 $
    Supplier Number: 12-6320
  • EaseAlign Evaporation cover, low actinic - red, with DDM opening, for 705-DS, VK7000,7010
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 224,00 $
    224,00 $
    Each
       224,00 $
    Supplier Number: 12-6311
  • Evaporation cover, for 100 ml tube, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 423,00 $
    423,00 $
    Each
       423,00 $
    Supplier Number: 27-1510
  • Evaporation cover, for use with EaseAlign, solid cover for non-vessel positions
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 60,10 $
    60,10 $
    Each
       60,10 $
    Supplier Number: 12-6330
  • Evaporation cover, low-loss hinged, for EaseAlign centering rings
    Discontinued
    Supplier Number: -
  • Evaporation cover, low-loss hinged, for use with resident probes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 438,00 $
    438,00 $
    Each
       438,00 $
    Supplier Number: 12-6329
  • Evaporation cover, low-loss split hinge design
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 322,00 $
    322,00 $
    Each
       322,00 $
    Supplier Number: 12-6327
  • Evaporation cover, USP, for 300 ml tube, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 298,00 $
    298,00 $
    Each
       298,00 $
    Supplier Number: 27-1500