Well Plate Tubes for Bond Elut Certify Plates, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
Bond Elut Certify is a mixed-mode phase ideal for extraction of basic drugs from urine and blood.
- Mixed-mode sorbent bed
- Broad application range for aqueous extraction
- Bimodal, nonpolar, and strong cation exchange
The Bond Elut Certify cartridge utilizes a packed bed consisting of a nonpolar C8 sorbent and a strong cation-exchange (SCX) sorbent. Certify is most used to extract basic (cationic) drugs from urine and blood. It is also very effective for extracting a wide range of compounds from a diverse number of aqueous matrices.
Frequently Bought Together