Dissolution Enhancer Cells, Agilent Technologies | Avantor
Dissolution Enhancer Cells, Agilent Technologies
Dissolution Enhancer Cells, Agilent Technologies
Certificates
Dissolution Enhancer Cells, Agilent Technologies
Dissolution Enhancer Cells, Agilent Technologies
7 Options Available Below
Product Details & Documents
The Enhancer (immersion) Cell is a PTFE cell with adjustable volume and a retaining cap.
  • Ideal for testing ointments, creams, and gels
  • Available in several surface areas, including 0.5, 2.0, and 4.0 cm²
  • Used with traditional dissolution apparatus and a 200 ml flat-bottom vessel or 1 L vessel
  • Meets SUPAC-Semisolid Guidance
  • Alignment tools specific to the surface area are sold individually
The Enhancer (immersion) Cell is a PTFE cell with adjustable volume and a retaining cap to eliminate air pockets trapped against the skin or artificial membrane that provide the diffusion barrier.
An alignment tool, available in various sizes to match membrane surface area size, is used to hold the membrane in place while eliminating air pockets with the universal adjustment tool.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Adjustment Tool, Enhancer Cell, fits all sizes, for 708-DS;705-DS;VK7000;VK7030
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $316.00
    $316.00
    Each
       $316.00
    Supplier Number: 12-4015
  • Alignment tool/Height gauge, for 2 cm. sq. Enhancer Cell, for 708-DS;705-DS;VK7000;VK7030
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $298.00
    $298.00
    Each
       $298.00
    Supplier Number: 12-4021
  • Alignment tool/Height gauge, for 4 cm. sq. Enhancer Cell, for 708-DS;705-DS;VK7000;VK7030
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $298.00
    $298.00
    Each
       $298.00
    Supplier Number: 12-4020
  • Enhancer Cell Kit 4 cm. sq., for 708-DS;705-DS;VK7000;VK7030
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $1,750.00
    $1,750.00
    Kit of 1
       $1,750.00
    Supplier Number: 12-4005
  • Enhancer Cell only, 2 cm. sq., for 708-DS;705-DS;VK7000;VK7030
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,400.00
    $1,400.00
    Each
       $1,400.00
    Supplier Number: 12-4001
  • Enhancer Cell only, 4 cm. sq., for 708-DS;705-DS;VK7000;VK7030
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,400.00
    $1,400.00
    Each
       $1,400.00
    Supplier Number: 12-4000
  • TruAlign vessel, clear glass, 200 ml, Flat Bottom, for Enhancer Cell, for 708-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $760.00
    $760.00
    Each
       $760.00
    Supplier Number: 12-5170