Dissolution Conversion Kits, Agilent Technologies | Avantor
Dissolution Conversion Kits, Agilent Technologies
Dissolution Conversion Kits, Agilent Technologies
Certificates
Dissolution Conversion Kits, Agilent Technologies
Dissolution Conversion Kits, Agilent Technologies
3 Options Available Below
Product Details & Documents
Agilent can also support small-volume dissolution with the 708-DS dissolution apparatus.
  • Special conversion kits: available in sets of 6 and 8, also available for the Chinese Pharmacopeia (CP) 250 ml vessel
  • Conversion kits are available to upgrade your 705-DS and 7000 and 7010 models to TruCenter vessels
The Agilent 708-DS dissolution apparatus can also support small-volume dissolution. Each position will require a conversion kit to hold a 100 or 200 ml vessel in position. The conversion kits are designed to be used with either mini paddles, mini baskets, or enhancer cells.
Easy-to-install conversion kits: the 708-DS is designed to accept 100 and 200 ml vessels, the 250 ml Chinese Pharmacopoeia vessel, and standard 1 L vessels. The 2 L model can incorporate 1 and 2 L vessels as well as smaller volume vessels with conversion kits.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Conversion kit, TruCenter, with ground groove, 1000 ml, for 705-DS and VK7000/7010
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $896.00
    $896.00
    Each
       $896.00
    Supplier Number: 12-6100
  • Conversion Kit, from 300 ml outer tubes to 1000 ml, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $17,870.00
    $17,870.00
    Kit of 1
       $17,870.00
    Supplier Number: 27-6105
  • TruAlign Vessel Conversion Kit, 100/200ml, for 708-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $795.00
    $795.00
    Each
       $795.00
    Supplier Number: 12-6368