Dissolution Centering Rings, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Dissolution Centering Rings, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
Jump to:
À propos de cet article
Centering rings are available in various types and sizes.
- Centering rings are available for the EaseAlign vessels of the legacy 705-DS, 7000, and 7010 dissolution apparatus
The correct positioning of vessels is critical for dissolution testing. Some dissolution vessels are held in place by a centering ring, which are available in various types and sizes.
2 Options Available Below
Product Details & Documents
Centering rings are available in various types and sizes.
- Centering rings are available for the EaseAlign vessels of the legacy 705-DS, 7000, and 7010 dissolution apparatus
The correct positioning of vessels is critical for dissolution testing. Some dissolution vessels are held in place by a centering ring, which are available in various types and sizes.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 12-6050
- Supplier Number: 12-6065
- TruCenter Alignment PlateDiscontinued-Supplier Number: 12-6115