CTC Smart Autosampler Syringes, Agilent Technologies
CTC Smart Autosampler Syringes, Agilent Technologies
CTC Smart Autosampler Syringes, Agilent Technologies
Certificates
CTC Smart Autosampler Syringes, Agilent Technologies
CTC Smart Autosampler Syringes, Agilent Technologies
15 Options Available Below
Product Details & Documents
Agilent now offers Smart Syringes compatible with your PAL3 Autosampler!
  • Smart technology enables critical parameter tracking for each unique syringe
  • Compatible with all PAL3 models
  • CTC syringes provide precise and effective sample delivery from your CTC autosampler to Agilent GC
  • Handles both liquid and headspace injections
Agilent now offers smart syringes compatible with your PAL3 Autosampler! Each smart syringe is equipped with its own read/write chip that allow you to track critical parameters, such as the number of injections a specific syringe has made, the max temperature it’s experienced, and more! Furthermore, these smart syringes allow your PAL3 instrument to automatically recognize and upload these parameters, for efficient and detailed tracking. For precise, effective, sample delivery from your CTC autosampler, Agilent has the right syringe for you. Whether you're performing liquid or headspace injections on your PAL3 autosampler, you'll find what you need among our wide range of syringes for CTC autosamplers.
Product Family Options
Product Information
  • Désignation
    Volume
    Needle Gauge
    Needle Length
    Needle Type
    Tip Type
    Availability
    Price
  • Smart Syringe, PAL3
    1 µl
    23
    57 mm
    Cone
    Log in for availability
    Log in for availability
    Each
    Each - $549.00
    $549.00
    Each
       $549.00
    Supplier Number: 8010-1343
  • Smart Syringe, PAL3
    10 µl
    23s
    57 mm
    Fixed
    Cone
    Log in for availability
    Log in for availability
    Each
    Each - $274.00
    $274.00
    Each
       $274.00
    Supplier Number: 8010-1303
  • Smart Syringe, PAL3
    10 µl
    26s
    57 mm
    Fixed
    Cone
    Log in for availability
    Log in for availability
    Each
    Each - $274.00
    $274.00
    Each
       $274.00
    Supplier Number: 8010-1304
  • Smart ITEX Syringe
    1300 µl (1.3 ml)
    Log in for availability
    Log in for availability
    Each
    Each - $768.00
    $768.00
    Each
       $768.00
    Supplier Number: 8010-1336
  • Smart GFHS Syringe, SP, PTFE tip plunger
    2250 µl (2.25 ml)
    23
    23 mm
    Fixed
    Side hole
    Log in for availability
    Log in for availability
    Each
    Each - $768.00
    $768.00
    Each
       $768.00
    Supplier Number: 8010-1340
  • Smart HS Syringe, PTFE tip plunger
    2500 µl (2.5 ml)
    23
    65 mm
    Fixed
    Side hole
    Log in for availability
    Log in for availability
    Each
    Each - $512.00
    $512.00
    Each
       $512.00
    Supplier Number: 8010-1338
  • Smart HS Syringe, PTFE tip plunger
    5000 µl (5 ml)
    23
    65 mm
    Fixed
    Side hole
    Log in for availability
    Log in for availability
    Each
    Each - $731.00
    $731.00
    Each
       $731.00
    Supplier Number: 8010-1339
  • Smart Syringe body, PTFE tip plunger
    10 µl
    Log in for availability
    Log in for availability
    Each
    Each - $183.00
    $183.00
    Each
       $183.00
    Supplier Number: 8010-1346
  • Smart Syringe body, PTFE tip plunger
    1000 µl (1 ml)
    Log in for availability
    Log in for availability
    Each
    Each - $274.00
    $274.00
    Each
       $274.00
    Supplier Number: 8010-1352
  • Smart Syringe, PAL3, PTFE tip plunger
    1000 µl (1 ml)
    22
    57 mm
    Fixed
    Flat
    Log in for availability
    Log in for availability
    Each
    Each - $329.00
    $329.00
    Each
       $329.00
    Supplier Number: 8010-1325
  • Smart Syringe, PAL3, PTFE tip plunger
    1000 µl (1 ml)
    23
    57 mm
    Fixed
    Cone
    Log in for availability
    Log in for availability
    Each
    Each - $402.00
    $402.00
    Each
       $402.00
    Supplier Number: 8010-1326
  • Smart Syringe, PAL3, PTFE tip plunger
    500 µl
    26
    57 mm
    Fixed
    Cone
    Discontinued
    Supplier Number: -
  • Smart Syringe, PAL3, PTFE tip plunger
    250 µl
    26
    85 mm
    Fixed
    Cone
    Log in for availability
    Log in for availability
    Each
    Each - $512.00
    $512.00
    Each
       $512.00
    Supplier Number: 8010-1321
  • Smart Syringe, PAL3, PTFE tip plunger
    100 µl
    22s
    57 mm
    Fixed
    Flat
    Log in for availability
    Log in for availability
    Each
    Each - $640.00
    $640.00
    Each
       $640.00
    Supplier Number: 8010-1319
  • Smart Syringe, PAL3, PTFE tip plunger
    10 µl
    23s
    57 mm
    Fixed
    Cone
    Log in for availability
    Log in for availability
    Each
    Each - $384.00
    $384.00
    Each
       $384.00
    Supplier Number: 8010-1306
  • Smart Syringe, PAL3, PTFE tip plunger
    100 µl
    23s
    57 mm
    Fixed
    Cone
    Log in for availability
    Log in for availability
    Each
    Each - $347.00
    $347.00
    Each
       $347.00
    Supplier Number: 8010-1316
Replacement Needles
Product Information
  • Catalog #
    Désignation
    Longueur
    Point Style
    Jauge
    Availability
    Price
  • Needle, stainless steel, for 10 µl syringe
    57.0 mm (2.24")
    Cone
    23s
    Log in for availability
    Log in for availability
    Each
    Each - $329.00
    $329.00
    Each
       $329.00
Product Information
  • Catalog #
    Désignation
    Longueur
    Point Style
    Jauge
    Availability
    Price
  • Needle, stainless steel, for 1 ml syringe
    57.0 mm (2.24")
    Side Hole
    23
    Log in for availability
    Log in for availability
    Each
    Each - $329.00
    $329.00
    Each
       $329.00
Product Information
  • Catalog #
    Désignation
    Longueur
    Point Style
    Jauge
    Availability
    Price
  • Needle, stainless steel, for 10 µl syringe
    56.0 mm (2.20")
    Cone
    26s
    Log in for availability
    Log in for availability
    Each
    Each - $329.00
    $329.00
    Each
       $329.00