- Description:Lens cleaning paper, lint free, 50/pack
- Cat. no.:CA76515-124
Agilent offers a highly effective alkaline cleaning concentrate for glass and quartz glass cuvettes.
- Highly effective alkaline concentrate and low phosphate content
- Significantly reduces the surface tension of water
- Removal of dirt particles is also assured by the good wetting action
- Concentrate is filtered to 1 μm and is therefore virtually free of solid particles
- Using demineralised water improves the cleaning characteristics
- Warming the cleaning solution can speed up the cleaning process
- Care is required to avoid thermal shock to the cuvettes
Regularly cleaning UV cuvettes or other optical components with cleaning solution ensures accurate measurement and precise results. Agilent offers a highly effective alkaline cleaning concentrate that is ideal for use on glass and quartz glass cuvettes, and other sensitive optical parts. Optimal dilution (usually 0.5 to 2 vol%) depends on factors such as the hardness of the water, the degree and type of contamination, temperature, etc.
The high emulsifying and dispersing capabilities prevent the redeposition of the loosened particles. Special surface-active substances facilitate the residue-free rinsing of the optical components once they have been cleaned.
Suitable for use with glass, quartz, sapphire, porcelain, ceramics, plastics, and ferrous metals (when diluted to a working concentration of 2%).