QuickSwap Restrictors for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
QuickSwap interface restrictors can increase the productivity of your 5973N and 5975 inert MSD systems, allowing you to change columns without venting the MSD.
- Prefabricated for convenience and ease of use
- For applications requiring other restrictor sizes, Agilent offers a variety of deactivated fused silica tubing, SilTitle ferrules and swaging tools
Agilent QuickSwap capillary flow technology module and preswaged, fused silica tubing restrictors can increase the productivity of your Agilent 5973N and 5975 inert MSD systems, allowing you to change columns without venting the MSD.
Frequently Bought Together