ContactUsLinkComponent
Cavity Chemical Injector, Mortech
Cavity Chemical Injector, Mortech
Catalog # 76549-778
undefined
Cavity Chemical Injector, Mortech
Catalog # 76549-778
Supplier Number:  FC001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    160 mm (6.25")
  • Cat. no.:
    76549-778
Specifications
About this item
The use of this injector for cavity fluids minimizes the fumes when injecting the cavities. It is leakproof under all conditions and fits any standard threaded fluid bottle.
  • The use of this injector for cavity fluids
  • Minimizes the fumes when injecting the cavities
  • It is leakproof under all conditions and fits any standard threaded fluid bottle