Biopsy Bags, Mortech | Avantor
Biopsy Bags, Mortech
Supplier:   Mopec
Biopsy Bags, Mortech
Supplier:  Mopec
Certificates
Biopsy Bags, Mortech
Biopsy Bags, Mortech
5 Options Available Below
Product Details & Documents
Easy to open and available in three convenient sizes, you’ll find our nylon mesh biopsy bags are the optimal solution for protecting specimens during tissue processing.
  • Ensure unrestricted fluid movement around tissue
  • Made from nylon, 0.2 mm mesh dia.
  • All seams are heat-sealed
  • Reduce reagent carryover and pressure artifacts often found with sponges
  • Rapidly remove specimen with convenient peel-open bag
  • Reduces the loss of small biopsy specimens
Available in standard or premium grade.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Standard
    Small
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $785.52
    $785.52
    Each
       $785.52
    Supplier Number: BI001
  • Standard
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $292.34
    $292.34
    Each
       $292.34
    Supplier Number: BI002
  • Standard
    Large
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $823.66
    $823.66
    Each
       $823.66
    Supplier Number: BI003
  • Premium
    Small
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,426.09
    $1,426.09
    Each
       $1,426.09
    Supplier Number: BI004
  • Premium
    Medium
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $602.73
    $602.73
    Each
       $602.73
    Supplier Number: BI005