Mopec Amplify Setting Solution, Mortech
Catalogue # 76549-704
Fournisseur: Mortech Manufacturing
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Size:250 ml (8 oz.) bottle
- Cat. no.:76549-704
Spécifications
À propos de cet article
Amplify is a fixative spray usually applied after the application of Harmony dyes, although it can also be applied to tissue prior to marking with dyes.
- Modifies the surface of challenging specimens
- Aids the adhesion of Harmony Tissue Marking Dyes
- Can be applied to tissue before its marked with dye
- Suitable for fatty specimens
The fixative spray modifies the surface of challenging specimens to aid the adhesion of Harmony dyes. It is strongly recommended to use Ink Fix for use with fatty specimens.