Paraffin Tapper Set, Mortech | Avantor
Paraffin Tapper Set, Mortech
Catalog # 76549-698
Supplier:   Mopec
Fournisseur #:  BG028
Paraffin Tapper Set, Mortech
Catalog # 76549-698
Supplier:  Mopec
Paraffin Tapper Set, Mortech
Paraffin Tapper Set, Mortech
Price
$655.96
Log in pour la disponibilité
Each
Product Details & Documents
Multiple sizes to fit different embedding molds and designed to flatten tissue during the embedding process.
  • Stainless steel storage base
Delivery information: Includes 5×5×1.5 mm paraffin tapper; 13×12×1.5 mm paraffin tapper; 22×22×1.5 mm paraffin tapper; 29×23×1.5 mm paraffin tapper; and 35×23×1.5 mm paraffin tapper.